Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013179.1 | NP_001013197.1 | 3 | 469 | ||
XM_006245407.2 | XP_006245469.1 | 4 | 422 | |||
GenBank mRNA | BC087597.1 | - | 3 | 469 | 7311379 |