Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013631.2 | NP_001013653.1 | 1, 2 | 68 | ||
GenBank mRNA | BC002696.2 | - | 1, 2 | 37 | ||
BC137258.1 | - | 1, 2 | 68 | 9021255 | ||
BX283835.1 | - | 1, 2 | 38 |
Gene Symbol: HNRNPCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136561.3 | NP_001130033.3 | 1, 2 | 54 | ||
GenBank mRNA | BC144551.1 | - | 1, 2 | 54 | 9053078 |