Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metrnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014104.1 | NP_001014126.1 | 2 | 473 | ||
GenBank mRNA | AB646250.1 | - | 3 | 226 | ||
BC086590.1 | - | 2 | 473 | 7310123 |