Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014108.1 | NP_001014130.1 | 5 | 1467 | ||
XM_006257580.2 | XP_006257642.1 | 6 | 1065 | |||
GenBank mRNA | BC081962.1 | - | 5 | 1467 | 7124728 |