Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014110.1 | NP_001014132.1 | 6, 7 | 830 | ||
GenBank mRNA | BC085918.1 | - | 6, 7 | 830 | 7108577 |