Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024712.2 | NP_001019883.2 | 90 | |||
GenBank mRNA | BC055874.1 | - | 89 | 3491523 |
Gene Symbol: Gm10409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY739387.1 | - | 97 |
Gene Symbol: Gm3020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012594.1 | - | 1, 2 | 113 | ||
BY711739.1 | - | 1, 2 | 113 |
Gene Symbol: Gm3500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256886.1 | NP_001243815.1 | 1, 2 | 115 |