Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025013.1 | NP_001020184.1 | 3 | 758 | ||
XM_006256976.1 | XP_006257038.1 | 4 | 818 | |||
XM_006256977.2 | XP_006257039.1 | 5 | 881 | |||
GenBank mRNA | BC079208.1 | - | 3 | 758 | 7131795 |