Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025103.2 | NP_001020274.2 | 10 | 1331 | ||
GenBank mRNA | BC026645.1 | - | 7 | 905 | ||
BC034124.1 | - | 4 | 527 | |||
BC048916.1 | - | 10 | 1331 | |||
BC098333.1 | - | 10 | 1292 | 5256663 |