Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC547349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025208.1 | NP_001020379.1 | 2 | 531 | ||
GenBank mRNA | AY533669.1 | - | 2 | 504 | ||
AY989869.1 | - | 2 | 531 |
Gene Symbol: H2-K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L23494.1 | - | 3 | 563 |