Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025284.1 | NP_001020455.1 | 7 | 1668 | ||
GenBank mRNA | BC098023.1 | - | 7 | 1668 | 7455606 |