Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 3 | 530 | ||
GenBank mRNA | AB062402.1 | - | 3 | 483 | ||
AF088851.1 | - | 3 | 337 | |||
AK311818.1 | - | 3 | 501 | |||
AY258285.1 | - | 1088 | ||||
BC000857.1 | - | 3 | 489 | 3459353 | ||
BC001399.2 | - | 3 | 458 | 3051458 | ||
BC011359.2 | - | 3 | 480 | 4157296 | ||
BC013724.2 | - | 3 | 483 | 3857790 | ||
BC015156.1 | - | 3 | 353 | 3883694 | ||
BC016009.1 | - | 3 | 505 | 4666545 | ||
BC016857.1 | - | 3 | 449 | 3848510 | ||
BC020300.1 | - | 3 | 466 | |||
BC035441.1 | - | 3 | 489 | |||
BC063514.1 | - | 3 | 472 | 3882345 | ||
BC066961.1 | - | 3 | 503 | 4796312 | ||
BC070494.1 | - | 4 | 444 | |||
BC073750.1 | - | 3 | 487 | 6256153 | ||
BC104643.1 | - | 3 | 505 | 4043391 | ||
BC105802.1 | - | 4 | 482 | |||
BQ887222.1 | - | 530 | ||||
L20941.1 | - | 3 | 503 | |||
M11146.1 | - | 3 | 372 | |||
M12937.1 | - | 3 | 321 | |||
M97164.1 | - | 3 | 424 | |||
X00318.1 | - | 3 | 386 |