Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003577.2 | NP_003568.2 | 1 | 470 | ||
GenBank mRNA | AB011076.2 | - | 1 | 471 | ||
AY606112.1 | - | 1 | 470 |