Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199653.1 | NP_001186582.1 | 5 | 590 | ||
NM_001199654.1 | NP_001186583.1 | 5 | 657 | |||
NM_007221.3 | NP_009152.2 | 5 | 651 | |||
GenBank mRNA | AF141310.1 | - | 5 | 614 | ||
AK124646.1 | - | 5 | 585 | |||
AK289490.1 | - | 5 | 632 | |||
AK290260.1 | - | 5 | 632 | |||
AL080101.1 | - | 5 | 620 | |||
BC050735.1 | - | 5 | 606 | |||
BC056417.1 | - | 5 | 615 | |||
BC065031.1 | - | 5 | 630 | 6148250 |