Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032532.2 | NP_115921.2 | 13 | 4285 | ||
XM_011536190.2 | XP_011534492.1 | 12 | 4208 | |||
XM_011536191.2 | XP_011534493.1 | 10 | 3926 | |||
GenBank mRNA | AB058769.2 | - | 11 | 3771 | ||
AJ586132.1 | - | 3 | 205 | |||
AK127635.1 | - | 3 | 293 | |||
AL832410.1 | - | 3 | 502 | |||
BC146783.1 | - | 11 | 3771 | 9007239 | ||
BC150607.1 | - | 12 | 3960 | 9056977 | ||
DQ009660.1 | - | 13 | 4285 |