Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032532.2 | NP_115921.2 | 23 | 6233 | ||
XM_011536190.2 | XP_011534492.1 | 22 | 6156 | |||
XM_011536191.2 | XP_011534493.1 | 20 | 5874 | |||
GenBank mRNA | AB058769.2 | - | 21 | 5701 | ||
AJ586132.1 | - | 13 | 2134 | |||
AK127635.1 | - | 13 | 2223 | |||
AL832410.1 | - | 13 | 2432 | |||
BC146783.1 | - | 21 | 5701 | 9007239 | ||
DQ009660.1 | - | 23 | 6215 |