Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032538.1 | NP_115927.1 | 15 | 5159 | ||
XM_011514947.2 | XP_011513249.1 | 14 | 4976 | |||
XM_011514948.2 | XP_011513250.1 | 3 | 5300 | |||
XM_017011364.1 | XP_016866853.1 | 13 | 5213 | |||
XM_017011365.1 | XP_016866854.1 | 13 | 5210 | |||
XM_017011366.1 | XP_016866855.1 | 14 | 4952 | |||
XM_017011368.1 | XP_016866857.1 | 3 | 5270 | |||
GenBank mRNA | AB058758.1 | - | 13 | 4728 | ||
AB218664.1 | - | 16 | 4963 | |||
AK055425.1 | - | 1 | 278 | |||
AL833256.1 | - | 1 | 64 | |||
BC036764.1 | - | 2 | 1951 |