Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159596.1 | NP_001153068.1 | 5 | 847 | ||
NM_017693.3 | NP_060163.2 | 7 | 1531 | |||
GenBank mRNA | AF339783.1 | - | 2 | 122 | ||
AF339793.1 | - | 3 | 920 | |||
AF411385.1 | - | 7 | 1531 | |||
AK000166.1 | - | 5 | 743 | |||
AK225229.1 | - | 5 | 743 | |||
AK290658.1 | - | 7 | 1400 | |||
AK304391.1 | - | 6 | 1285 | |||
BC020663.1 | - | 5 | 675 | |||
BC051813.2 | - | 5 | 656 | |||
BC075084.2 | - | 5 | 879 | 30915598 | ||
BC075085.2 | - | 5 | 879 | 30915386 | ||
BC112339.1 | - | 5 | 602 | 40053977 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 5 | 974 | ||
GenBank mRNA | AB209312.1 | - | 5 | 766 | ||
LM994822.1 | - | 5 | 852 |