Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164060.1 | NP_001157532.1 | 1 | 666 | ||
NM_053517.2 | NP_445969.2 | 2 | 233 | |||
GenBank mRNA | BC088298.1 | - | 2 | 233 | 7323045 | |
D83015.1 | - | 1 | 14 |