Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134360.1 | NP_599187.1 | 8, 9 | 700 | ||
XM_008762464.1 | XP_008760686.1 | 8, 9 | 756 | |||
XM_008762465.1 | XP_008760687.1 | 7, 8 | 700 | |||
GenBank mRNA | BC097949.1 | - | 8, 9 | 700 | 7387440 |