Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMF1-BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199661.1 | NP_001186590.1 | 7 | 766 | ||
NM_001199662.1 | NP_001186591.1 | 7 | 827 | |||
NM_001199663.1 | NP_001186592.1 | 6 | 631 | |||
NM_001199664.1 | NP_001186593.1 | 5 | 620 | |||
GenBank mRNA | BC033656.2 | - | 6 | 621 | ||
FY213424.1 | - | 5 | 533 |
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199173.5 | NP_954642.1 | 4 | 569 | ||
GenBank mRNA | BC113432.1 | - | 4 | 371 | 8322484 | |
BC113434.1 | - | 4 | 371 | 8322486 | ||
X51699.1 | - | 4 | 317 | |||
X53698.1 | - | 4 | 317 |