Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000197.1 | - | 9 | 1872 | ||
AY424283.1 | - | 9 | 1872 | |||
BC039234.1 | - | 10 | 1582 | 4608045 |