Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000443.1 | NP_001000443.1 | 496 |
Gene Symbol: LOC100910863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766088.1 | XP_008764310.1 | 1 | 496 |
Gene Symbol: LOC100911801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762665.1 | XP_008760887.1 | 1 | 496 |