Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171191.1 | NP_001164662.1 | 10, 11 | 1459 | ||
NM_001171193.1 | NP_001164664.1 | 11, 12 | 1563 | |||
NM_017711.3 | NP_060181.2 | 12, 13 | 1677 | |||
XM_011530977.1 | XP_011529279.1 | 9, 10 | 1193 | |||
XM_017029614.1 | XP_016885103.1 | 11, 12 | 1658 | |||
GenBank mRNA | AB048363.1 | - | 12, 13 | 1567 | ||
AK000214.1 | - | 12, 13 | 1388 | |||
AK097293.1 | - | 7, 8 | 1155 | |||
AK299255.1 | - | 10, 11 | 1287 | |||
AK310090.1 | - | 7, 8 | 1517 | |||
AK316035.1 | - | 11, 12 | 1295 | |||
AY358986.1 | - | 12, 13 | 1372 | |||
BC032009.1 | - | 12, 13 | 1374 | 4747934 |