Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 53 | 6221 | ||
NM_014689.2 | NP_055504.2 | 53 | 6364 | |||
XM_005246673.4 | XP_005246730.1 | 53 | 6223 | |||
XM_006712617.3 | XP_006712680.1 | 53 | 6388 | |||
XM_017004441.1 | XP_016859930.1 | 53 | 6391 | |||
XM_017004442.1 | XP_016859931.1 | 53 | 6367 | |||
XM_017004443.1 | XP_016859932.1 | 53 | 6440 | |||
XM_017004444.1 | XP_016859933.1 | 52 | 6347 | |||
XM_017004445.1 | XP_016859934.1 | 53 | 6221 | |||
XM_017004446.1 | XP_016859935.1 | 52 | 6245 | |||
XM_017004447.1 | XP_016859936.1 | 53 | 6363 | |||
GenBank mRNA | AB074179.1 | - | 8 | 954 | ||
AF250396.1 | - | 13 | 1640 | |||
AK000227.1 | - | 11 | 1333 | |||
AK001253.1 | - | 8 | 964 | |||
AK293847.1 | - | 19 | 2290 | |||
AK293879.1 | - | 24 | 2677 | |||
AK307914.1 | - | 13 | 1725 | |||
BC015018.1 | - | 11 | 1331 | |||
CR749492.1 | - | 28 | 3209 | |||
EU236710.2 | - | 53 | 6218 |