Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001513.2 | NP_001001513.2 | 6, 7 | 793 | ||
XM_006246745.2 | XP_006246807.1 | 6, 7 | 1036 | |||
GenBank mRNA | AY607588.1 | - | 6, 7 | 500 | ||
BC079107.1 | - | 6, 7 | 793 | 7125293 | ||
FQ221626.1 | - | 6, 7 | 422 |