Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002791.2 | NP_001002791.1 | 2 | 255 | ||
GenBank mRNA | AK078980.1 | - | 2 | 256 | ||
BC139230.1 | - | 2 | 256 | 8862252 | ||
BC145928.1 | - | 2 | 256 | 40131231 | ||
DQ012033.1 | - | 2 | 255 |