Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFWT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002916.4 | NP_001002916.3 | 1 | 368 | ||
GenBank mRNA | AY283370.1 | - | 1 | 341 | ||
BC038109.2 | - | 1 | 365 | 5266336 | ||
BC118604.1 | - | 1 | 361 | 40109007 | ||
BC121816.1 | - | 1 | 361 | 40109006 |
Gene Symbol: H2BFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164416.1 | NP_001157888.1 | 1 | 306 | ||
XM_006724640.2 | XP_006724703.1 | 1 | 339 | |||
XM_011530922.2 | XP_011529224.1 | 1 | 502 | |||
GenBank mRNA | AK093522.1 | - | 1 | 306 |