Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003841.2 | NP_001003841.1 | 3, 4 | 531 | ||
GenBank mRNA | AK096054.1 | - | 3, 4 | 439 | ||
AK290811.1 | - | 3, 4 | 531 | |||
AY591756.1 | - | 3, 4 | 475 | |||
AY596807.1 | - | 3, 4 | 475 |