Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004076.2 | NP_001004076.2 | 6 | 701 | ||
XM_006250626.2 | XP_006250688.1 | 6 | 750 | |||
XM_008770002.1 | XP_008768224.1 | 6 | 3091 | |||
XM_008770003.1 | XP_008768225.1 | 7 | 2971 | |||
GenBank mRNA | AY688113.1 | - | 5 | 628 | ||
BC098896.1 | - | 6 | 701 | 7100556 |
Gene Symbol: Abcg3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107205.1 | NP_001100675.1 | 6 | 686 | ||
XM_006250606.2 | XP_006250668.1 | 6 | 1325 |