Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 510 | ||
XM_017001691.1 | XP_016857180.1 | 3 | 415 | |||
GenBank mRNA | AF151042.1 | - | 3 | 494 | ||
AF374386.1 | - | 3 | 428 | |||
AK000319.1 | - | 2 | 231 | |||
BC001959.1 | - | 3 | 481 | 3536977 | ||
BC078177.1 | - | 3 | 462 | 6526331 | ||
BC085018.1 | - | 3 | 480 | 6650445 | ||
DA043474.1 | - | 3 | 520 |