Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004721.1 | NP_001004721.1 | 7 | 605 | ||
GenBank mRNA | AK143517.1 | - | 7 | 605 | ||
AK162913.1 | - | 580 | ||||
AK171988.1 | - | 7 | 584 | |||
AW046539.1 | - | 2 | 86 | |||
BC120680.1 | - | 7 | 594 | 40129603 | ||
BC120682.1 | - | 7 | 594 | 40129605 | ||
CK334159.1 | - | 7 | 558 |