Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017772.3 | NP_060242.2 | 10 | 1336 | ||
NR_130108.1 | 10 | 1397 | ||||
XM_011514738.2 | XP_011513040.1 | 10 | 1288 | |||
XM_011514739.1 | XP_011513041.1 | 10 | 1288 | |||
GenBank mRNA | AB449909.1 | - | 10 | 1122 | ||
AK000344.1 | - | 3 | 243 | |||
AK096340.1 | - | 10 | 1266 | |||
AK292893.1 | - | 10 | 1231 | |||
AY313781.1 | - | 10 | 1268 | |||
AY927466.1 | - | 3 | 417 | |||
AY927467.1 | - | 3 | 424 | |||
BC002720.2 | - | 3 | 196 | |||
BC047658.1 | - | 10 | 1329 | |||
BC063523.1 | - | 5 | 346 | |||
BC109026.1 | - | 10 | 1266 | 40027698 | ||
BC109027.1 | - | 10 | 1266 | 40027700 | ||
BX648317.1 | - | 10 | 1267 | |||
CR749710.1 | - | 4 | 422 |