Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114120.2 | NP_001107592.1 | 2 | 309 | ||
NM_017779.5 | NP_060249.2 | 2 | 309 | |||
GenBank mRNA | AB281187.1 | - | 2 | 165 | ||
AB281274.1 | - | 2 | 165 | |||
AJ278112.1 | - | 2 | 165 | |||
AK000361.1 | - | 2 | 170 | |||
AK225735.1 | - | 2 | 155 | |||
BC003511.1 | - | 2 | 158 | |||
BC065304.1 | - | 2 | 112 | 6156084 | ||
DB147703.1 | - | 2 | 309 |