Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017785.4 | NP_060255.3 | 3 | 599 | ||
XM_011534585.1 | XP_011532887.1 | 3 | 545 | |||
XM_011534586.2 | XP_011532888.1 | 3 | 481 | |||
XM_011534587.2 | XP_011532889.1 | 4 | 746 | |||
XM_011534588.2 | XP_011532890.1 | 4 | 684 | |||
XM_011534589.2 | XP_011532891.1 | 3 | 494 | |||
XM_011534590.2 | XP_011532892.1 | 2 | 235 | |||
XM_017009601.1 | XP_016865090.1 | 3 | 430 | |||
XM_017009602.1 | XP_016865091.1 | 2 | 235 | |||
GenBank mRNA | AF269167.1 | - | 3 | 490 | ||
AK000371.1 | - | 2 | 325 | |||
AK308835.1 | - | 3 | 478 | |||
AK310442.1 | - | 3 | 398 | |||
AY517557.1 | - | 3 | 430 | |||
BC012568.1 | - | 3 | 465 | 4102558 |