Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 901 | |||
NR_027791.3 | 6 | 704 | ||||
NR_111004.2 | 6 | 770 | ||||
NR_111005.2 | 7 | 831 | ||||
NR_136542.1 | 8 | 965 | ||||
NR_136543.1 | 5 | 643 | ||||
NR_136544.1 | 4 | 856 | ||||
NR_136545.1 | 5 | 971 | ||||
NR_136546.1 | 3 | 795 | ||||
NR_136547.1 | 3 | 406 | ||||
NR_136548.1 | 4 | 660 | ||||
NR_136549.1 | 4 | 587 | ||||
NR_136550.1 | 4 | 595 | ||||
NR_136551.1 | 6 | 739 | ||||
NR_136552.1 | 7 | 854 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 901 |