Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000812.3 | NP_000803.2 | 2 | 521 | ||
XM_017007986.1 | XP_016863475.1 | 2 | 246 | |||
GenBank mRNA | AK225167.1 | - | 2 | 198 | ||
AK225686.1 | - | 2 | 225 | |||
AK293691.1 | - | 2 | 166 | |||
AK296023.1 | - | 2 | 414 | |||
AK307120.1 | - | 2 | 248 | |||
AK312720.1 | - | 2 | 167 | |||
BC022449.1 | - | 2 | 225 | 4797401 | ||
DC352362.1 | - | 2 | 521 | |||
KJ534841.1 | - | 2 | 147 | |||
KJ535054.1 | - | 2 | 147 | |||
X14767.1 | - | 2 | 178 |