Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008708.2 | NP_001008708.1 | 3 | 678 | ||
GenBank mRNA | BC017941.1 | - | 3 | 651 | 4282697 | |
BC019239.1 | - | 3 | 682 | 4050858 | ||
BC025376.1 | - | 3 | 682 | 4045703 | ||
BC053896.1 | - | 3 | 655 | 6174351 |