Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-CE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008838.1 | NP_001008838.1 | 1 | 94 |
Gene Symbol: LOC103694386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772782.1 | XP_008771004.1 | 2 | 259 |