Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008904.1 | NP_001008904.1 | 1 | 317 | ||
GenBank mRNA | AY510277.1 | - | 1 | 317 |
Gene Symbol: LOC102551504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228159.1 | XP_006228221.1 | 1 | 317 |