Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009708.1 | NP_001009708.1 | 1145 | |||
XM_006233438.2 | XP_006233500.1 | 911 | ||||
GenBank mRNA | BC087700.1 | - | 1145 | 7314152 | ||
FQ233103.1 | - | 989 |