Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009714.1 | NP_001009714.1 | 476 | |||
XM_008767036.1 | XP_008765258.1 | 550 | ||||
GenBank mRNA | BC088263.1 | - | 476 | 7376620 |