Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009831.2 | NP_001009831.1 | 5 | 726 | ||
XM_006240177.1 | XP_006240239.1 | 4 | 662 | |||
GenBank mRNA | AY581896.2 | - | 5 | 726 | ||
BC092645.1 | - | 4 | 649 | 7318365 |