Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010898.2 | NP_001010898.1 | 12 | 4667 | ||
GenBank mRNA | AK124875.1 | - | 1 | 1972 | ||
AL833249.1 | - | 1 | 515 |