Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM206A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017832.3 | NP_060302.1 | 2 | 190 | ||
XM_011518816.2 | XP_011517118.1 | 2 | 186 | |||
GenBank mRNA | AF153417.1 | - | 2 | 120 | ||
AK000464.1 | - | 2 | 174 | |||
AK310297.1 | - | 2 | 146 | |||
BC015795.1 | - | 2 | 402 | 3910513 |