Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142363.1 | NP_001135835.1 | 6 | 839 | ||
XM_006248558.2 | XP_006248620.1 | 6 | 857 | |||
GenBank mRNA | BC088475.1 | - | 5 | 722 | 7129249 | |
FM075565.1 | - | 5 | 651 |