Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012161.1 | NP_001012161.1 | 8, 9 | 815 | ||
XM_006254015.2 | XP_006254077.1 | 8, 9 | 871 | |||
XM_008771736.1 | XP_008769958.1 | 8, 9 | 787 | |||
GenBank mRNA | BC089833.1 | - | 8, 9 | 815 | 7325371 |