Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012161.1 | NP_001012161.1 | 15, 16 | 1480 | ||
XM_006254015.2 | XP_006254077.1 | 15, 16 | 1536 | |||
XM_008771736.1 | XP_008769958.1 | 15, 16 | 1452 | |||
GenBank mRNA | BC089833.1 | - | 15, 16 | 1480 | 7325371 | |
FQ234296.1 | - | 7, 8 | 605 |