Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012164.1 | NP_001012164.1 | 17 | 2169 | ||
XM_006255278.2 | XP_006255340.1 | 17 | 2836 | |||
XM_006255279.2 | XP_006255341.1 | 16 | 2701 | |||
XM_006255280.1 | XP_006255342.1 | 18 | 2418 | |||
XM_006255281.2 | XP_006255343.1 | 15 | 2554 | |||
GenBank mRNA | BC083857.1 | - | 17 | 2169 | 7125718 |
Set Membership: |
G-protein coupled receptor |