Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012167.1 | NP_001012167.1 | 10 | 1241 | ||
XM_006228578.2 | XP_006228640.1 | 12 | 1570 | |||
XM_006228579.2 | XP_006228641.1 | 12 | 1518 | |||
XM_006228580.2 | XP_006228642.1 | 11 | 1676 | |||
XM_006228581.2 | XP_006228643.1 | 12 | 1548 | |||
XM_008759165.1 | XP_008757387.1 | 11 | 1461 | |||
GenBank mRNA | BC089987.1 | - | 10 | 1241 | 7318612 |