Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000924.3 | NP_000915.1 | 3 | 563 | ||
NM_001165975.2 | NP_001159447.1 | 2 | 251 | |||
NM_001288768.1 | NP_001275697.1 | 3 | 563 | |||
NM_001288769.1 | NP_001275698.1 | 2 | 183 | |||
NM_001315534.1 | NP_001302463.1 | 2 | 251 | |||
NM_001315535.1 | NP_001302464.1 | 2 | 251 | |||
XM_011538456.1 | XP_011536758.1 | 4 | 513 | |||
XM_017019432.1 | XP_016874921.1 | 2 | 82 | |||
XM_017019433.1 | XP_016874922.1 | 2 | 770 | |||
GenBank mRNA | AB209200.1 | - | 3 | 332 | ||
AJ401609.1 | - | 2 | 110 | |||
AK126899.1 | - | 2 | 194 | |||
AK296422.1 | - | 2 | 183 | |||
AK302899.1 | - | 3 | 290 | |||
AK302931.1 | - | 2 | 238 | |||
BC032226.1 | - | 3 | 336 | 5200139 | ||
BP342896.1 | - | 2 | 230 | |||
BX538092.1 | - | 2 | 251 | |||
DC298452.1 | - | 2 | 380 | |||
DC392024.1 | - | 2 | 238 | |||
EU668337.1 | - | 3 | 332 | |||
U56976.1 | - | 2 | 204 | |||
U86078.1 | - | 3 | 283 |